Assay Details
| Gene Symbol: Numbl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010950.2 | NP_035080.2 | 5 - 6 | 575 | 82 | |
| GenBank mRNA | AK147523.1 | - | 3 - 4 | 6362 | 82 | |
| BC059915.1 | - | 4 - 5 | 1164 | 82 | ||
| BC068116.1 | - | 5 - 6 | 402 | 6809443 | 82 | |
| BC098097.1 | - | 5 - 6 | 416 | 6403012 | 82 | |
| U96441.1 | - | 5 - 6 | 459 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Numbl
Genomic Map