Assay Details
| Gene Symbol: Numbl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010950.2 | NP_035080.2 | 7 - 8 | 909 | 142 | |
| GenBank mRNA | AK147523.1 | - | 5 - 6 | 6696 | 142 | |
| BC059915.1 | - | 6 - 7 | 1498 | 142 | ||
| BC068116.1 | - | 7 - 8 | 736 | 6809443 | 142 | |
| BC098097.1 | - | 7 - 8 | 750 | 6403012 | 142 | |
More Information
Set Membership: |
|

Numbl
Genomic Map