Assay Details
| Gene Symbol: Numbl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010950.2 | NP_035080.2 | 8 - 9 | 1211 | 56 | |
| GenBank mRNA | AK147523.1 | - | 6 - 7 | 6998 | 56 | |
| BC059915.1 | - | 7 - 8 | 1800 | 56 | ||
| BC068116.1 | - | 8 - 9 | 1038 | 6809443 | 56 | |
| BC098097.1 | - | 8 - 9 | 1052 | 6403012 | 56 | |
| U96441.1 | - | 9 - 10 | 1092 | 56 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Numbl
Genomic Map