Assay Details
| Gene Symbol: Aplp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007467.3 | NP_031493.2 | 4 - 5 | 647 | 71 | |
| GenBank mRNA | AK154992.1 | - | 4 - 5 | 601 | 71 | |
| AK160871.1 | - | 4 - 5 | 599 | 71 | ||
| AK170818.1 | - | 4 - 5 | 580 | 71 | ||
| BC021877.1 | - | 4 - 5 | 626 | 4481838 | 71 | |
| BC106158.1 | - | 3 - 4 | 735 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Aplp1
Genomic Map