Assay Details
| Gene Symbol: Aplp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007467.3 | NP_031493.2 | 6 - 7 | 980 | 72 | |
| GenBank mRNA | AK160871.1 | - | 6 - 7 | 932 | 72 | |
| AK170818.1 | - | 6 - 7 | 913 | 72 | ||
| BC021877.1 | - | 6 - 7 | 959 | 4481838 | 72 | |
| BC106158.1 | - | 5 - 6 | 1068 | 72 | ||
| L04538.1 | - | 6 - 7 | 950 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Aplp1
Genomic Map