Assay Details
| Gene Symbol: Arhgap33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289670.1 | NP_001276599.1 | 14 - 15 | 1438 | 70 | |
| NM_001289682.1 | NP_001276611.1 | 15 - 16 | 1521 | 70 | ||
| NM_178252.3 | NP_839983.1 | 15 - 16 | 1548 | 70 | ||
| GenBank mRNA | AK142180.1 | - | 14 - 15 | 1439 | 70 | |
| AY217764.1 | - | 14 - 15 | 1381 | 70 | ||
| BC061471.1 | - | 8 - 9 | 725 | 5698780 | 70 | |
| BC065086.1 | - | 15 - 16 | 1497 | 6850475 | 70 | |
| BC065166.1 | - | 15 - 16 | 1495 | 6836510 | 70 | |
| BC066047.1 | - | 15 - 16 | 1473 | 6822191 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arhgap33
Genomic Map