Assay Details
| Gene Symbol: Map2k1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008927.3 | NP_032953.1 | 9 - 10 | 1328 | 72 | |
| GenBank mRNA | AB041567.1 | - | 9 - 10 | 1308 | 72 | |
| AK146931.1 | - | 9 - 10 | 1321 | 72 | ||
| AK154793.1 | - | 9 - 10 | 1355 | 72 | ||
| AK165664.1 | - | 8 - 9 | 862 | 72 | ||
| AK165894.1 | - | 9 - 10 | 1329 | 72 | ||
| AK167336.1 | - | 9 - 10 | 1295 | 72 | ||
| AK192210.1 | - | 4 - 5 | 380 | 72 | ||
| BC051137.1 | - | 9 - 10 | 1147 | 72 | ||
| BC054754.1 | - | 9 - 10 | 1107 | 6411664 | 72 | |
| L02526.1 | - | 9 - 10 | 1055 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Map2k1
Genomic Map