Assay Details
| Gene Symbol: Tipin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025372.3 | NP_079648.1 | 4 - 5 | 333 | 88 | |
| GenBank mRNA | AK003451.1 | - | 4 - 5 | 327 | 88 | |
| AK011357.1 | - | 4 - 5 | 328 | 88 | ||
| AK038111.1 | - | 332 | 88 | |||
| BC016211.1 | - | 3 - 4 | 310 | 4924605 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tipin
Genomic Map