Assay Details
| Gene Symbol: Adam10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007399.3 | NP_031425.2 | 11 - 12 | 1717 | 113 | |
| GenBank mRNA | AF011379.1 | - | 11 - 12 | 1538 | 113 | |
| AK036599.1 | - | 5 - 6 | 761 | 113 | ||
| AK036883.1 | - | 3 - 4 | 405 | 113 | ||
| AK036918.1 | - | 3 - 4 | 406 | 113 | ||
| BC057346.1 | - | 11 - 12 | 1678 | 113 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adam10
Genomic Map