Assay Details
| Gene Symbol: Mto1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026658.2 | NP_080934.2 | 5 - 6 | 1023 | 88 | |
| GenBank mRNA | AF369902.1 | - | 5 - 6 | 1013 | 88 | |
| AK019088.1 | - | 5 - 6 | 1023 | 88 | ||
| BC063256.1 | - | 5 - 6 | 1009 | 88 | ||
| CF173757.1 | - | 4 - 5 | 558 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mto1
Genomic Map