Assay Details
| Gene Symbol: Mto1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026658.2 | NP_080934.2 | 7 - 8 | 1347 | 64 | |
| GenBank mRNA | AF369902.1 | - | 7 - 8 | 1337 | 64 | |
| AK019088.1 | - | 7 - 8 | 1347 | 64 | ||
| AK188313.1 | - | 1 - 2 | 63 | 64 | ||
| AK210384.1 | - | 1 - 2 | 63 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mto1
Genomic Map