Assay Details
| Gene Symbol: Mto1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026658.2 | NP_080934.2 | 8 - 9 | 1550 | 61 | |
| GenBank mRNA | AF369902.1 | - | 8 - 9 | 1540 | 61 | |
| AK019088.1 | - | 8 - 9 | 1550 | 61 | ||
| BC063256.1 | - | 8 - 9 | 1547 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mto1
Genomic Map