Assay Details
| Gene Symbol: Ddx43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191044.1 | NP_001177973.1 | 8 - 9 | 1141 | 148 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ddx43| Gene Symbol: Ddx43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191044.1 | NP_001177973.1 | 8 - 9 | 1141 | 148 | |
Set Membership: |
|