Assay Details
| Gene Symbol: Olfr948 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011756.1 | NP_001011756.1 | 1 | 906 | 82 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P477972 | NM_001011756.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P477972 | NM_001011756.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 395620 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P477972 | NM_001011756.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P477972 | NM_001011756.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P477972 | NM_001011756.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P477972 | NM_001011756.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26438090 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26438078 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26438086 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26438091 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26438054 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26438066 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26438072 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26438071 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26438087 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26438074 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26438060 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26438089 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26438063 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26438044 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26438070 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26438077 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26438068 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26438079 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26438052 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26438069 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26438080 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26438085 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26438067 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26438051 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26438082 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26438038 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26438058 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26438042 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26438040 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26438057 | - | - |
| Whole Mouse Genome G4122A | A_51_P477972 | NM_001011756.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P477972 | NM_001011756.1 | mouse_transcriptome |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08037019 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07739568 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08037020 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08037021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26438071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26438095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26438044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26438068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26438074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26438082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26438069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26438052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26438100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26438070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26438101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26438093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26438090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26438078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26438089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26438113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26438063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26438115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26438105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26438098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26438112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26438060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26438085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26438054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26438042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26438080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26438087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26438066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26438058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26438099 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26438082 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26438079 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26438086 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26438039 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26438101 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26438085 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08037021 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26438074 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26438054 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26438063 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26438060 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26438057 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26438100 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26438066 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26438078 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26438070 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26438080 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26438071 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26438069 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26438095 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26438087 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07857520 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26438052 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26438098 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26438044 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26438089 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26438038 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26438096 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26438099 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26438040 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26438042 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26438058 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26438090 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07858100 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26438068 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07705090 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07638095 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10592405 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10592404 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900017196.mm.1 | NM_001011756.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900017196.mm.1 | NM_001011756.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900017196.mm.1 | TC0900002075.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 355332 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1250401 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2804649 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2804649 | - | - |

Olfr948
Genomic Map