Assay Details
| Gene Symbol: 4833428L15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040732.1 | 1 - 2 | 88 | 84 | ||
| GenBank mRNA | AK014786.1 | - | 1 - 2 | 90 | 84 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Down syndrome cell adhesion molecule like 1 | Dscaml1 |
4921507G06Rik, 4930435C18Rik, BB239540, mKIAA1132 |
Anti-sense | Chr. 9 - 45429495 - 45753713 on NCBI Build GRCm38 | |
| Downstream | centrosomal protein 164 | Cep164 |
AI450905, BC027092, D030051D21, mKIAA1052 |
335.7kb | Sense | Chr. 9 - 45766946 - 45828686 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

4833428L15Rik
Genomic Map