Assay Details
| Gene Symbol: Fam103a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025997.2 | NP_080273.1 | 2 - 3 | 301 | 95 | |
| GenBank mRNA | AK010694.1 | - | 2 - 3 | 257 | 95 | |
| AK011881.1 | - | 2 - 3 | 301 | 95 | ||
| AK018614.1 | - | 2 - 3 | 300 | 95 | ||
| AK079819.1 | - | 2 - 3 | 280 | 95 | ||
| AK081301.1 | - | 2 - 3 | 293 | 95 | ||
| AK085230.1 | - | 2 - 3 | 299 | 95 | ||
| AK150938.1 | - | 2 - 3 | 268 | 95 | ||
| AK169392.1 | - | 2 - 3 | 330 | 95 | ||
| BC058367.1 | - | 2 - 3 | 224 | 6830048 | 95 | |
| BC091760.1 | - | 2 - 3 | 261 | 5042613 | 95 | |
| BC096399.1 | - | 2 - 3 | 259 | 876882 | 95 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25273846 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25273838 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25273843 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25273837 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25273842 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25273844 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25273836 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25273845 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25273839 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25273848 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2032267 | NM_001177750.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2032267 | AK139034.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2085127 | BC096399.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2085127 | AK160347.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2085127 | AK169392.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2085127 | BC058367.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2085127 | BC091760.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 706809 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 475642 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 739601 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 504070 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 109460 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P168836 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2085127 | BC096399.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2085127 | AK160347.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2085127 | AK169392.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2085127 | BC058367.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2085127 | BC091760.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25273834 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25273847 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25273827 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25273853 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25273849 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25273832 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25273837 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25273830 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25273843 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25273854 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25273850 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25273829 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25273840 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25273838 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25273855 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25273825 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25273842 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P168836 | - | - |
| Whole Mouse Genome G4122A | A_51_P127958 | BC096399.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P127958 | AK160347.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P127958 | AK169392.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P127958 | BC058367.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P127958 | BC091760.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P330766 | AK160347.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P330766 | AK169392.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P330766 | BC058367.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P330766 | AK079819.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P330766 | AK081301.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P102188 | BC096399.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P102188 | AK160347.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P102188 | AK169392.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P102188 | BC091760.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P102188 | AK081301.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P113125 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P168836 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06129829 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06129820 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06511030 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06129822 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06511027 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25273846 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25273837 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25273845 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25273836 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25273848 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25273839 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25273838 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25273844 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25273842 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25273843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25273918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25273901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25273865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25273859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25273827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25273916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25273835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25273922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25273837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25273894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25273897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25273933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25273847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25273872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25273891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25273925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25273923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25273914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25273854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25273840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25273893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25273920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25273821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25273907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25273862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25273903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25273934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25273829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25273909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25273820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25273849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25273932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25273915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25273885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25273818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25273856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25273887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25273851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25273863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25273834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25273911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25273832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25273830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25273889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25273855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25273853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25273842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25273899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25273921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25273905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25273824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25273825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25273867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25273861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25273838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25273930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25273870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25273860 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25273847 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25273860 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25273829 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25273835 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05926390 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25273851 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25273864 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25273845 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06404167 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06511027 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25273849 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06511028 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25273899 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25273883 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06511030 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06129828 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25273853 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06314007 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25273842 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25273863 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25273870 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25273827 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25273850 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25273837 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25273894 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25273885 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25273830 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25273867 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25273865 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25273854 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25273872 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25273903 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05996086 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25273900 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25273862 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25273825 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25273840 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25273901 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06554480 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25273897 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06314008 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25273855 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25273859 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25273889 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06404168 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06020714 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25273843 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25273887 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10584759 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10554572 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10554571 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10554570 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10554573 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1428966_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10584758 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10554569 | - | - |
| Mouse Genome 430 2.0 Array | 1428966_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1428966_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700010386.mm.1 | TC0700001177.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700010386.mm.1 | NM_025997.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700010386.mm.1 | NM_025997.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700010388.mm.1 | TC0700001177.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700010388.mm.1 | NM_025997.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700010388.mm.1 | NM_025997.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700010389.mm.1 | TC0700001177.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700010389.mm.1 | NM_025997.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700010389.mm.1 | NM_025997.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700010379.mm.1 | TC0700001177.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700010379.mm.1 | NM_025997.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700010379.mm.1 | NM_025997.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700010381.mm.1 | TC0700001177.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700010381.mm.1 | NM_025997.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700010381.mm.1 | NM_025997.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700010387.mm.1 | TC0700001177.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700010387.mm.1 | NM_025997.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700010387.mm.1 | NM_025997.2 | Refseq |
| Mouse Expression Set 430 | 1428966_at | - | - |
| Mouse Expression Set 430 | 1428966_PM_at | - | - |
| Murine Genome U74v2 Set | 93059_PM_at | - | - |
| Murine Genome U74v2 Set | 93059_at | - | - |
| Mouse Genome 430A 2.0 Array | 1428966_at | - | - |
| Mouse Genome 430A 2.0 Array | 1428966_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 899480 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2850711 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002850711 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1219307 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1219307 | - | - |
| mouse-6 v1.1 (Illumina) | scl067148.6_4-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2850711 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl067148.6_4-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2850711 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2810443 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2810443 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl067148.6_4-S | - | - |

Fam103a1
Genomic Map