Assay Details
| Gene Symbol: Sin3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110350.1 | NP_001103820.1 | 8 - 9 | 1605 | 86 | |
| NM_001110351.1 | NP_001103821.1 | 9 - 10 | 1482 | 86 | ||
| NM_011378.2 | NP_035508.2 | 9 - 10 | 1713 | 86 | ||
| GenBank mRNA | AK032209.1 | - | 8 - 9 | 1473 | 86 | |
| AK136347.1 | - | 3 - 4 | 407 | 86 | ||
| AK220292.1 | - | 8 - 9 | 1582 | 86 | ||
| BC052716.1 | - | 8 - 9 | 1492 | 6837144 | 86 | |
| BC053385.1 | - | 8 - 9 | 1475 | 86 | ||
| BC078454.1 | - | 8 - 9 | 1466 | 86 | ||
| L36831.1 | - | 8 - 9 | 1396 | 86 | ||
| L38620.2 | - | 7 - 8 | 1372 | 86 | ||
| L38621.2 | - | 7 - 8 | 1372 | 86 | ||
| U22394.1 | - | 9 - 10 | 1721 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sin3a
Genomic Map