Assay Details
| Gene Symbol: 4930407I19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK019573.1 | - | 4 - 5 | 754 | 98 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 1700023F02 gene | 1700023F02Rik | AV205046 | -49.1kb | Anti-sense | Chr. 10 - 66120609 - 66124064 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 1110002J07 gene | 1110002J07Rik | 732.7kb | Anti-sense | Chr. 10 - 66912191 - 66920616 on NCBI Build GRCm38 |
More Information
Set Membership: |
|
MicroArray Probes:
Agilent
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2004906 | AK019573.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 577303 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 9727 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 927315 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 533503 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 827944 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 330251 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 142390 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 636575 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 659381 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 414925 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 288541 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 106631 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2004906 | AK019573.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07088870 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07088893 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P105092 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08969942 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205331 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205347 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205294 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205508 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205290 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205471 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205424 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205485 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205299 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08365316 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205342 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205506 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205488 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205509 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08220176 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205499 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205441 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205542 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205423 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205398 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205367 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205501 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205322 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08942972 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205417 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205300 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205302 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205480 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205404 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08918146 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205376 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205316 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205551 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08383042 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205359 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205319 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205504 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08931806 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08222070 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205397 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205391 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205337 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205309 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205422 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205413 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205335 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08363139 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205420 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205371 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205548 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205438 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205533 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205356 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205312 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205393 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205374 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205402 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205384 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205486 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205400 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205340 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205432 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205472 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08767815 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205306 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205487 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205483 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205292 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205388 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205326 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205538 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205436 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08239075 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205304 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205540 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205547 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205440 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205536 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205365 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205368 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205387 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205500 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205383 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205556 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205297 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205439 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08327044 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205534 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205295 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205377 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205489 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205546 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205395 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205433 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205466 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205313 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205380 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205360 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205555 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205361 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205305 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205358 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205435 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205363 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205554 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205467 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205419 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205344 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205333 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08293357 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08663818 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205550 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205343 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205470 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205329 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08942974 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205443 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205430 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08325311 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205415 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205303 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08663817 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27205382 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08396979 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08834285 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 101770154 | - | - |

4930407I19Rik
Genomic Map