Assay Details
| Gene Symbol: Gm30853 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK076946.1 | - | 4 - 5 | 337 | 85 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
catenin (cadherin associated protein), alpha 3 |
Ctnna3 |
4930429L08Rik, 4933408A16, Catna3, Vr22 |
-877.9kb | Anti-sense | Chr. 10 - 63430098 - 65005395 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 1700023F02 gene | 1700023F02Rik | AV205046 | 227.0kb | Sense | Chr. 10 - 66120609 - 66124064 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm30853
Genomic Map