Assay Details
| Gene Symbol: Pip5k1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293647.1 | NP_001280576.1 | 16 - 17 | 2014 | 81 | |
| NM_008844.3 | NP_032870.2 | 16 - 17 | 2001 | 81 | ||
| GenBank mRNA | AB006916.1 | - | 16 - 17 | 1984 | 81 | |
| BC094665.1 | - | 16 - 17 | 1973 | 30356181 | 81 | |
More Information
Set Membership: |
|

Pip5k1c
Genomic Map