Assay Details
| Gene Symbol: Amh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007445.2 | NP_031471.2 | 3 - 4 | 659 | 64 | |
| GenBank mRNA | AY911505.1 | - | 3 - 4 | 659 | 64 | |
| BC150154.1 | - | 3 - 4 | 658 | 64 | ||
| BC150477.1 | - | 3 - 4 | 658 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Amh
Genomic Map