Assay Details
| Gene Symbol: Mknk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB164081.1 | - | 9 - 10 | 750 | 126 | |
| AK030830.1 | - | 4 - 5 | 302 | 126 | ||
| AK154235.1 | - | 10 - 11 | 1030 | 126 | ||
| AK194740.1 | - | 3 - 4 | 187 | 126 | ||
| AK194893.1 | - | 3 - 4 | 187 | 126 | ||
| BC010256.1 | - | 8 - 9 | 660 | 3257646 | 126 | |
| Y11092.1 | - | 8 - 9 | 701 | 126 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mknk2
Genomic Map