Assay Details
| Gene Symbol: Mknk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB164081.1 | - | 3 - 4 | 243 | 75 | |
| AK154235.1 | - | 4 - 5 | 523 | 75 | ||
| BC010256.1 | - | 2 - 3 | 153 | 3257646 | 75 | |
| BY732164.1 | - | 3 - 4 | 246 | 75 | ||
| KF612275.1 | - | 3 - 4 | 243 | 75 | ||
| Y11092.1 | - | 2 - 3 | 194 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mknk2
Genomic Map