Assay Details
| Gene Symbol: Mfsd12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028657.3 | NP_082933.2 | 6 - 7 | 1036 | 95 | |
| GenBank mRNA | AK149678.1 | - | 6 - 7 | 1042 | 95 | |
| AK154386.1 | - | 6 - 7 | 1038 | 95 | ||
| AK154902.1 | - | 6 - 7 | 1116 | 95 | ||
| AK170185.1 | - | 6 - 7 | 1056 | 95 | ||
| AK171084.1 | - | 6 - 7 | 1038 | 95 | ||
| BC118620.1 | - | 6 - 7 | 1036 | 40107417 | 95 | |
| BC119790.1 | - | 6 - 7 | 1036 | 40107416 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mfsd12
Genomic Map