Assay Details
| Gene Symbol: Fzr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019757.1 | NP_062731.1 | 8 - 9 | 861 | 94 | |
| GenBank mRNA | AF083809.1 | - | 8 - 9 | 861 | 94 | |
| AK051993.1 | - | 8 - 9 | 944 | 94 | ||
| AK081476.1 | - | 8 - 9 | 916 | 94 | ||
| AK154822.1 | - | 8 - 9 | 945 | 94 | ||
| AK155370.1 | - | 8 - 9 | 943 | 94 | ||
| AK163951.1 | - | 8 - 9 | 920 | 94 | ||
| BC006616.1 | - | 8 - 9 | 942 | 3967009 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fzr1
Genomic Map