Assay Details
| Gene Symbol: Dapk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190474.1 | NP_001177403.2 | 1 - 2 | 93 | 106 | |
| GenBank mRNA | AK158299.1 | - | 1 - 2 | 84 | 106 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001190474.1 transcripts.
Set Membership: |
|

Dapk3
Genomic Map