Assay Details
| Gene Symbol: Dapk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190473.1 | NP_001177402.1 | 3 - 4 | 505 | 63 | |
| NM_001190474.1 | NP_001177403.2 | 3 - 4 | 557 | 63 | ||
| NM_007828.2 | NP_031854.1 | 2 - 3 | 525 | 63 | ||
| GenBank mRNA | AB007143.1 | - | 2 - 3 | 438 | 63 | |
| AK035171.1 | - | 1 - 2 | 127 | 63 | ||
| AK158299.1 | - | 3 - 4 | 548 | 63 | ||
| BC012210.1 | - | 3 - 4 | 523 | 63 | ||
| BC125443.1 | - | 2 - 3 | 525 | 40129958 | 63 | |
| BC137680.1 | - | 2 - 3 | 525 | 8860699 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dapk3
Genomic Map