Assay Details
| Gene Symbol: Cactin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027381.2 | NP_081657.1 | 5 - 6 | 1337 | 91 | |
| GenBank mRNA | AK010963.1 | - | 2 - 3 | 146 | 91 | |
| AY934504.1 | - | 5 - 6 | 1243 | 91 | ||
| BC023696.1 | - | 2 - 3 | 257 | 91 | ||
| BC150986.1 | - | 5 - 6 | 1337 | 9087899 | 91 | |
| BC151180.1 | - | 5 - 6 | 1337 | 9088069 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of protein phosphorylation
immune system process
mRNA processing
multicellular organism development
RNA splicing
negative regulation of lipopolysaccharide-mediated signaling pathway
negative regulation of NF-kappaB transcription factor activity
negative regulation of interferon-beta production
negative regulation of interleukin-8 production
negative regulation of tumor necrosis factor production
negative regulation of toll-like receptor signaling pathway
innate immune response
negative regulation of type I interferon-mediated signaling pathway
cellular response to lipopolysaccharide
cellular response to interleukin-1
cellular response to tumor necrosis factor
poly(A) RNA binding
negative regulation of protein phosphorylation
immune system process
mRNA processing
multicellular organism development
RNA splicing
negative regulation of lipopolysaccharide-mediated signaling pathway
negative regulation of NF-kappaB transcription factor activity
negative regulation of interferon-beta production
negative regulation of interleukin-8 production
negative regulation of tumor necrosis factor production
negative regulation of toll-like receptor signaling pathway
innate immune response
negative regulation of type I interferon-mediated signaling pathway
cellular response to lipopolysaccharide
cellular response to interleukin-1
cellular response to tumor necrosis factor
poly(A) RNA binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27288890 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27288887 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27288894 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27288863 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27288891 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27288853 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27288892 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27288854 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27288886 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27288855 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27288893 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27288849 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27288889 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27288850 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27288864 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27288851 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27288895 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27288888 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27288856 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27288847 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P281806 | BY651766.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P281806 | AY934504.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P281806 | BC023696.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P281806 | AK010963.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P281806 | NM_027381.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P281806 | BY651766.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P281806 | AY934504.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P281806 | BC023696.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P281806 | AK010963.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P281806 | NM_027381.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 615464 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 728956 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 588883 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 139320 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P131674 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P281806 | BY651766.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P281806 | AY934504.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P281806 | BC023696.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P281806 | AK010963.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P281806 | NM_027381.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P281806 | BY651766.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P281806 | AY934504.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P281806 | BC023696.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P281806 | AK010963.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P281806 | NM_027381.2 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P131674 | - | - |
| Whole Mouse Genome G4122A | A_51_P281806 | BY651766.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P281806 | AY934504.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P281806 | BC023696.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P281806 | AK010963.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P281806 | NM_027381.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P281806 | BY651766.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P281806 | AY934504.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P281806 | BC023696.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P281806 | AK010963.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P281806 | NM_027381.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P482849 | BC151180.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P482849 | AY934504.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P482849 | BC023696.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P482849 | BC150986.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P482849 | AK010963.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P131674 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27288889 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27288887 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27288895 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27288894 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27288851 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27288855 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27288893 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27288888 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27288864 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27288863 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27288892 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27288886 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27288847 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27288850 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27288890 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27288856 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27288854 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27288849 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27288853 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27288891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288892 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08893478 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27288878 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27288843 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27288866 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27288861 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08949982 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27288831 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27288835 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27288890 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27288859 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27288875 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27288871 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27288882 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27288888 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27288832 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27288897 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27288849 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27288874 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08397161 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08397144 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27288876 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27288838 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27288845 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08694554 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08397158 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08294539 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27288872 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27288839 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08893479 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08694558 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27288885 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27288892 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27288830 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27288840 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27288846 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08397138 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10365096 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10365089 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10365095 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10365094 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10365097 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10365090 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10365091 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10365093 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10365092 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1452760_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10365088 | - | - |
| Mouse Genome 430 2.0 Array | 1452760_at | - | - |
| Mouse Genome 430 2.0 Array | 1452760_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 70312_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 70312_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1000007040.mm.1 | NM_027381.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000007040.mm.1 | NM_027381.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000007040.mm.1 | TC1000000893.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000007043.mm.1 | NM_027381.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000007043.mm.1 | NM_027381.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000007043.mm.1 | TC1000000893.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000007039.mm.1 | NM_027381.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000007039.mm.1 | NM_027381.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000007039.mm.1 | TC1000000893.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000007042.mm.1 | NM_027381.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000007042.mm.1 | NM_027381.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000007042.mm.1 | TC1000000893.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000007041.mm.1 | NM_027381.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000007041.mm.1 | NM_027381.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000007041.mm.1 | TC1000000893.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1452760_PM_at | - | - |
| Mouse Expression Set 430 | 1452760_at | - | - |
| Murine Genome U74v2 Set | 128770_PM_f_at | - | - |
| Murine Genome U74v2 Set | 167815_f_at | - | - |
| Murine Genome U74v2 Set | 128770_f_at | - | - |
| Murine Genome U74v2 Set | 167815_PM_f_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1452760_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1452760_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 838398 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2429710 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1255323 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2974280 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1255323 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2974280 | - | - |

Cactin
Genomic Map