Assay Details
| Gene Symbol: Creb3l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145365.3 | NP_663340.1 | 6 - 7 | 984 | 75 | |
| GenBank mRNA | AF392874.1 | - | 4 - 5 | 635 | 75 | |
| AK050186.1 | - | 4 - 5 | 626 | 75 | ||
| AK077258.1 | - | 6 - 7 | 985 | 75 | ||
| BC010786.1 | - | 4 - 5 | 679 | 4211480 | 75 | |
| BC028820.1 | - | 4 - 5 | 656 | 5096773 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Creb3l3
Genomic Map