Assay Details
| Gene Symbol: Sh3rf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172788.3 | NP_766376.2 | 2 - 3 | 1047 | 73 | |
| GenBank mRNA | AK029202.1 | - | 2 - 3 | 950 | 73 | |
| AK133071.1 | - | 2 - 3 | 1047 | 73 | ||
| BC057304.1 | - | 2 - 3 | 303 | 6830567 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sh3rf3
Genomic Map