Assay Details
| Gene Symbol: Sh3rf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172788.3 | NP_766376.2 | 3 - 4 | 1142 | 91 | |
| GenBank mRNA | AK029202.1 | - | 3 - 4 | 1045 | 91 | |
| AK133071.1 | - | 3 - 4 | 1142 | 91 | ||
| BC057304.1 | - | 3 - 4 | 398 | 6830567 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sh3rf3
Genomic Map