Assay Details
| Gene Symbol: Hnrnpl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177301.5 | NP_796275.3 | 11 - 12 | 1631 | 86 | |
| GenBank mRNA | AB009392.1 | - | 4 - 5 | 450 | 86 | |
| AK006480.1 | - | 1 - 2 | 37 | 86 | ||
| BC027206.1 | - | 11 - 12 | 1608 | 4218471 | 86 | |
| BC030461.1 | - | 4 - 5 | 474 | 86 | ||
| BC099683.1 | - | 11 - 12 | 1626 | 6530863 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hnrnpl
Genomic Map