Assay Details
| Gene Symbol: Cadm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145977.1 | NP_001139449.1 | 4 - 5 | 1179 | 77 | |
| NM_178721.4 | NP_848836.2 | 5 - 6 | 1206 | 77 | ||
| GenBank mRNA | AK033973.1 | - | 4 - 5 | 806 | 77 | |
| AK038842.1 | - | 5 - 6 | 1185 | 77 | ||
| AK043760.1 | - | 5 - 6 | 1189 | 77 | ||
| AK046800.1 | - | 4 - 5 | 1082 | 77 | ||
| AK163636.1 | - | 5 - 6 | 1183 | 77 | ||
| AK165030.1 | - | 5 - 6 | 1203 | 77 | ||
| BC139401.1 | - | 5 - 6 | 775 | 8862423 | 77 | |
| BC139402.1 | - | 5 - 6 | 775 | 8862424 | 77 | |
More Information
Set Membership: |
|

Cadm2
Genomic Map