Assay Details
Gene Symbol: Zfp708 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012325.2 | NP_001012325.2 | 1 - 2 | 186 | 99 | |
NM_001012449.2 | NP_001012451.2 | 1 - 2 | 186 | 99 | ||
GenBank mRNA | BG080714.2 | - | 1 - 2 | 149 | 99 | |
CF174027.1 | - | 1 - 2 | 186 | 99 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001012325.2, NM_001012449.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
