Assay Details
| Gene Symbol: Atp1a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290469.1 | NP_001277398.1 | 1 - 2 | 181 | 68 | |
| GenBank mRNA | AK053751.1 | - | 1 - 2 | 153 | 68 | |
| BC020177.1 | - | 1 - 2 | 139 | 4506376 | 68 | |
| BC034645.1 | - | 1 - 2 | 137 | 5357731 | 68 | |
| BC037206.1 | - | 1 - 2 | 136 | 5362423 | 68 | |
| BC042894.1 | - | 1 - 2 | 181 | 5364812 | 68 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC020177.1, BC034645.1, BC037206.1, BC042894.1, NM_001290469.1 transcripts.
Set Membership: |
|

Atp1a3
Genomic Map