Assay Details
| Gene Symbol: Pip5k1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146687.2 | NP_001140159.1 | 16 - 17 | 2006 | 99 | |
| GenBank mRNA | AK014403.1 | - | 16 - 17 | 1972 | 99 | |
| AK041402.1 | - | 16 - 17 | 1992 | 99 | ||
| AK154816.1 | - | 16 - 17 | 1998 | 99 | ||
| AK155371.1 | - | 16 - 17 | 1982 | 99 | ||
| AK170286.1 | - | 16 - 17 | 1998 | 99 | ||
| AK171576.1 | - | 16 - 17 | 1980 | 99 | ||
| AK179190.1 | - | 2 - 3 | 168 | 98 | ||
| AK210765.1 | - | 2 - 3 | 168 | 99 | ||
| AK216154.1 | - | 2 - 3 | 168 | 99 | ||
| AK218899.1 | - | 2 - 3 | 168 | 99 | ||
| BC019138.1 | - | 16 - 17 | 1955 | 5041431 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pip5k1c
Genomic Map