Assay Details
| Gene Symbol: Ftcd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080845.2 | NP_543121.1 | 9 - 10 | 1154 | 74 | |
| GenBank mRNA | AK165414.1 | - | 7 - 8 | 1361 | 74 | |
| BC010813.1 | - | 9 - 10 | 1116 | 4211065 | 74 | |
| BC024078.1 | - | 9 - 10 | 1148 | 5098529 | 74 | |
| CB949427.1 | - | 3 - 4 | 292 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ftcd
Genomic Map