Assay Details
| Gene Symbol: Agpat3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK076414.1 | - | 1 - 2 | 102 | 57 | |
| AK159110.1 | - | 1 - 2 | 373 | 57 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK076414.1, AK159110.1 transcripts.
Set Membership: |
|

Agpat3
Genomic Map