Assay Details
| Gene Symbol: Fgf22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC119135.1 | - | 2 - 3 | 333 | 8733884 | 78 |
| BC144814.1 | - | 2 - 3 | 333 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fgf22
Genomic Map