Assay Details
| Gene Symbol: Slc1a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009200.3 | NP_033226.1 | 6 - 7 | 1114 | 152 | |
| GenBank mRNA | AK159197.1 | - | 6 - 7 | 1115 | 152 | |
| BC138120.1 | - | 6 - 7 | 1016 | 8861139 | 152 | |
| BC138140.1 | - | 6 - 7 | 1016 | 8861159 | 152 | |
| D83262.1 | - | 6 - 7 | 1108 | 152 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc1a6
Genomic Map