Assay Details
| Gene Symbol: Fam13c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143777.1 | NP_001137249.1 | 2 - 3 | 271 | 89 | |
| GenBank mRNA | AK032053.1 | - | 2 - 3 | 83 | 89 | |
| AK036299.1 | - | 2 - 3 | 234 | 89 | ||
| AK039513.1 | - | 2 - 3 | 235 | 89 | ||
| AK044049.1 | - | 2 - 3 | 194 | 89 | ||
| AK081939.1 | - | 2 - 3 | 272 | 89 | ||
| AK220421.1 | - | 2 - 3 | 107 | 89 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK036299.1, NM_001143777.1 transcripts.
Set Membership: |
|

Fam13c
Genomic Map