Assay Details
| Gene Symbol: Fam13c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143776.1 | NP_001137248.1 | 2 - 3 | 219 | 128 | |
| NM_001143777.1 | NP_001137249.1 | 3 - 4 | 323 | 128 | ||
| NM_024244.4 | NP_077206.3 | 2 - 3 | 219 | 128 | ||
| GenBank mRNA | AK004797.1 | - | 2 - 3 | 203 | 128 | |
| AK021133.1 | - | 3 - 4 | 162 | 128 | ||
| AK032053.1 | - | 3 - 4 | 135 | 128 | ||
| AK036299.1 | - | 3 - 4 | 286 | 128 | ||
| AK039513.1 | - | 3 - 4 | 287 | 128 | ||
| AK041147.1 | - | 2 - 3 | 220 | 128 | ||
| AK044049.1 | - | 3 - 4 | 246 | 128 | ||
| AK081939.1 | - | 3 - 4 | 324 | 128 | ||
| AK082202.1 | - | 2 - 3 | 215 | 128 | ||
| AK220421.1 | - | 3 - 4 | 159 | 128 | ||
| BC129839.1 | - | 2 - 3 | 175 | 40060810 | 128 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK036299.1, NM_001143777.1 transcripts.
Set Membership: |
|

Fam13c
Genomic Map