Assay Details
| Gene Symbol: Fam13c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143776.1 | NP_001137248.1 | 4 - 5 | 547 | 62 | |
| NM_001143777.1 | NP_001137249.1 | 5 - 6 | 651 | 62 | ||
| NM_024244.4 | NP_077206.3 | 4 - 5 | 547 | 62 | ||
| GenBank mRNA | AK004797.1 | - | 4 - 5 | 531 | 62 | |
| AK032053.1 | - | 5 - 6 | 463 | 62 | ||
| AK036299.1 | - | 5 - 6 | 614 | 62 | ||
| AK039513.1 | - | 5 - 6 | 615 | 62 | ||
| AK041147.1 | - | 4 - 5 | 548 | 62 | ||
| AK044049.1 | - | 5 - 6 | 574 | 62 | ||
| AK082202.1 | - | 4 - 5 | 543 | 62 | ||
| AK220421.1 | - | 5 - 6 | 487 | 62 | ||
| BC129839.1 | - | 4 - 5 | 503 | 40060810 | 62 | |
| BC129840.1 | - | 4 - 5 | 503 | 40060811 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam13c
Genomic Map