Assay Details
| Gene Symbol: Pcbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021568.2 | NP_067543.2 | 1 - 2 | 199 | 78 | |
| GenBank mRNA | AK034811.1 | - | 1 - 2 | 140 | 78 | |
| BY275026.1 | - | 1 - 2 | 199 | 78 | ||
| BY277145.1 | - | 1 - 2 | 199 | 78 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK034811.1, NM_021568.2 transcripts.
Set Membership: |
|

Pcbp3
Genomic Map