Assay Details
Gene Symbol: Olfr1353 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_147042.2 | NP_667253.1 | 3 | 1138 | 111 | |
GenBank mRNA | BC119112.1 | - | 1 | 887 | 8733861 | 111 |
BC120824.1 | - | 1 | 887 | 40129747 | 111 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P289279 | NM_147042.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P289279 | AF042361.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P289279 | BC119112.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P289279 | BC120824.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P289279 | NM_147042.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P289279 | AF042361.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P289279 | BC119112.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P289279 | BC120824.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P144349 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P289279 | NM_147042.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P289279 | AF042361.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P289279 | BC119112.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P289279 | BC120824.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P289279 | NM_147042.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P289279 | AF042361.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P289279 | BC119112.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P289279 | BC120824.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P289279 | NM_147042.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P289279 | AF042361.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P289279 | BC119112.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P289279 | BC120824.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P289279 | NM_147042.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P289279 | AF042361.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P289279 | BC119112.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P289279 | BC120824.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27276059 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27276061 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27276076 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27276088 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27276051 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27276057 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27276095 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27276055 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27276079 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27276070 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27276058 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27276075 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27276048 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27276053 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27276078 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27276072 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27276082 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27276056 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27276064 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27276073 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27276084 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27276071 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27276077 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27276069 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27276086 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27276093 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27276066 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27276063 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27276092 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27276091 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P144349 | - | - |
Whole Mouse Genome G4122A | A_51_P289279 | NM_147042.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P289279 | AF042361.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P289279 | BC119112.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P289279 | BC120824.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P289279 | NM_147042.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P289279 | AF042361.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P289279 | BC119112.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P289279 | BC120824.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P131834 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P144349 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08872695 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08872697 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08872701 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27276061 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27276050 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27276069 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27276063 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27276088 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27276104 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27276076 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27276057 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27276093 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27276053 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27276086 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27276079 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27276092 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27276055 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27276051 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27276071 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27276064 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27276084 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27276072 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27276075 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27276058 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27276059 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27276091 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27276082 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27276069 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27276084 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27276070 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27276063 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27276076 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27276066 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27276092 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27276061 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27276071 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27276093 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27276091 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27276057 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27276048 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27276088 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27276053 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08872697 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08898102 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08332929 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27276051 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08761414 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08872701 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27276077 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27276082 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27276086 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27276059 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27276064 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27276046 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27276072 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27276102 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27276096 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10364420 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10364422 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10364421 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10364419 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 498248 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 3170253 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 3170253 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0003170253 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2632327 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2632327 | - | - |
mouse-6 v1.1 (Illumina) | scl38719.1.1_243-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 3170253 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl38719.1.1_243-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3170253 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2632327 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2632327 | - | - |
