Assay Details
| Gene Symbol: Gm10024 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081452.2 | NP_001074921.1 | 1 | 311 | 64 | |
| GenBank mRNA | AI561920.1 | - | 1 | 64 | ||
| AI586068.1 | - | 1 | 64 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Gm10024
Genomic Map