| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Gene 1.0 ST Array |
10364330 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10364329 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10364328 |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR1000005627.mm.1 |
NM_001162944.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000005627.mm.1 |
NM_001162944.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000005627.mm.1 |
TC1000000769.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000005626.mm.1 |
NM_001162944.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000005626.mm.1 |
NM_001162944.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000005626.mm.1 |
TC1000000769.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000005625.mm.1 |
NM_001162944.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000005625.mm.1 |
NM_001162944.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000005625.mm.1 |
TC1000000769.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000005624.mm.1 |
NM_001162944.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000005624.mm.1 |
NM_001162944.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000005624.mm.1 |
TC1000000769.mm.1 |
MTA TC |