Assay Details
| Gene Symbol: Adora2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009630.3 | NP_033760.2 | 1 - 2 | 272 | 60 | |
| GenBank mRNA | BY168137.1 | - | 1 - 2 | 206 | 60 | |
| BY349524.1 | - | 1 - 2 | 272 | 60 | ||
| CJ135665.1 | - | 1 - 2 | 206 | 60 | ||
| CX241487.1 | - | 1 - 2 | 60 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_009630.3 transcripts.
Set Membership: |
|

Adora2a
Genomic Map