Assay Details
| Gene Symbol: Reep3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204915.1 | NP_001191844.1 | 5 - 6 | 590 | 85 | |
| NM_178606.5 | NP_848721.1 | 5 - 6 | 590 | 85 | ||
| GenBank mRNA | AK005026.1 | - | 5 - 6 | 536 | 85 | |
| AK008058.1 | - | 5 - 6 | 509 | 85 | ||
| AK012403.1 | - | 5 - 6 | 534 | 85 | ||
| AK028868.1 | - | 5 - 6 | 547 | 85 | ||
| AK047728.1 | - | 5 - 6 | 458 | 85 | ||
| AK079649.1 | - | 5 - 6 | 460 | 85 | ||
| AK154559.1 | - | 5 - 6 | 448 | 85 | ||
| AY562231.1 | - | 5 - 6 | 423 | 85 | ||
| BC004607.1 | - | 5 - 6 | 499 | 3585133 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Reep3
Genomic Map