Assay Details
| Gene Symbol: Reep3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178606.5 | NP_848721.1 | 7 - 8 | 877 | 86 | |
| GenBank mRNA | AK005026.1 | - | 7 - 8 | 823 | 86 | |
| AK008058.1 | - | 7 - 8 | 796 | 86 | ||
| AK028868.1 | - | 7 - 8 | 834 | 86 | ||
| AK047728.1 | - | 7 - 8 | 745 | 86 | ||
| AK079649.1 | - | 7 - 8 | 747 | 86 | ||
| AY562231.1 | - | 7 - 8 | 710 | 86 | ||
| BC004607.1 | - | 7 - 8 | 786 | 3585133 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Reep3
Genomic Map